Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Selenium Plasma Standards

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Seq:
    2
  • Environmentally Preferable:
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

About this item