Order Entry
Belgium
ContactUsLinkComponent
 
J.T.Baker® BAKER INSTRA-ANALYZED®, Mercury Plasma Standard
  5736.0100
 :  Avantor
 :  
J.T.Baker® BAKER INSTRA-ANALYZED®, Mercury Plasma Standard
  5736.0100
 :  Avantor
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg