Order Entry
Belgium
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Iron Plasma Standards
J.T.Baker® BAKER INSTRA-ANALYZED®, Iron Plasma Standards
Catalog # 5731.0100
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER INSTRA-ANALYZED®, Iron Plasma Standards
Catalog # 5731.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Seq:
    2
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item