Order Entry
Belgium
Orders LinkContactUsLinkComponent
STERI-BUFFER

STERI-BUFFER

Phosphate buffer solution, size: 99 ml (3,3 oz.), 72
Catalog # VELTSB100-99
Supplier:   VELTEK ASSOCIATES
STERI-BUFFER
Catalog # VELTSB100-99
Supplier:  VELTEK ASSOCIATES
CAS Number:  
STERI-BUFFER
STERI-BUFFER

Phosphate buffer solution, size: 99 ml (3,3 oz.), 72

CAS Number:  
Price
569,00 €
Log in for availability
1 * 72 ST
Product Details & Documents

Developed for use as a buffer solution and diluent in lab testing such a bioburden testing, pharmaceutical microbial limits testing, and AET testing and as a rinsing agent.

Sealed with a 'no tamper' strip. Filtered at 0,2 µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.

Documents

Specifications

Specifications

  • Pk:
    72
  • Description:
    Phosphate buffer solution, size: 99 ml (3,3 oz.)

Specifications