Order Entry
Belgium
ContactUsLinkComponent
Recombinant uridine phosphorylase Salmonella typhimurium
Recombinant uridine phosphorylase Salmonella typhimurium
Catalog # APOSBITP1358-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant uridine phosphorylase Salmonella typhimurium
Catalog # APOSBITP1358-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Enzyme type:
    Recombinant
  • Storage conditions:
    Ambient
  • Enzyme name:
    Uridine phosphorylase Salmonella typhimurium
  • Enzyme synonyms:
    UPase

Specifications

About this item