Recombinant collagenase G
Catalog # APOSBIPR1004-2000C
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1
- Enzyme type:Recombinant
- Storage conditions:Freezer
- MDL No.:MFCD00130830
- Enzyme name:Collagenase G
- Pk Info:2000CDU
Specifications
About this item
From Clostridium histolyticum and produced in E. Coli and B. Megaterium. Endotoxin negative. No detectable Caseinase activity. Sterile lyophilized powder.