Tritirachium album proteinase K
Supplier: Apollo Scientific
Certificates
About this item
Proteinase K is a stable and highly reactive serineprotease
Specifications
- Species:Tritirachium album
- Storage conditions:Freezer
- MDL No.:MFCD00132129
- Enzyme name:Proteinase K
- Enzyme synonyms:Endopeptidase K
Specifications
Frequently Bought Together