Order Entry
Belgium
ContactUsLinkComponent
Candida antarctica Lipase B
Candida antarctica Lipase B
Supplier:  Apollo Scientific
Certificates
Candida antarctica Lipase B
Supplier:  Apollo Scientific
Certificates

Specifications

  • Species:
    Candida antarctica
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Lipase B

Specifications

Related Information

Frequently Bought Together