Order Entry
Belgium
ContactUsLinkComponent
Anti-CD5 Mouse Antibody
Anti-CD5 Mouse Antibody
Catalog # APOSBITP2007-1MG
Supplier:  Apollo Scientific
Anti-CD5 Mouse Antibody
Catalog # APOSBITP2007-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen symbol:
    CD5
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Pk:
    1 mg

Specifications

About this item