HyClone™ SFX-Insect™ cell culture media
HyClone™ SFX-Insect™ with L-glutamine is a versatile protein-free cell culture medium developed through metabolic pathway design to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. It provides superior growth of many key insect cell lines, including Sf9, Sf21, and High Five (T.ni), requiring minimal adaptation.
- Metabolically designed for high cell yield and recombinant protein production
- Complexed lipids for enhanced stability
- Supports multiple insect cell lines
- Supports direct adaptation
- High performance growth in multiple culture systems including high density bioreactors
The media are formulated using a lipid and phosopholipid complexing process for enhanced stability and performance and contain 0,1% Pluronic® F-68 for superior performance. It has been successfully tested in a variety of culture systems ranging from T-Flasks, spinners, shaker flasks, and various bioreactors including wave and hollow fibre. Manufactured according to cGMP guidelines.
Frequently Bought Together