ContactUsLinkComponent
Decontaminant, DNA-ExitusPlus™ Indicator Free
Decontaminant, DNA-ExitusPlus™ Indicator Free
Catalog # APOSBIA7409-5X500
Supplier:  Apollo Scientific
CAS Number:  
Decontaminant, DNA-ExitusPlus™ Indicator Free
Catalog # APOSBIA7409-5X500
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Pack type:
    Spray bottle
  • Pk:
    5
  • Description:
    DNA-ExitusPlus™ Indicator Free, 5×500 ml
  • Seq:
    04
  • Environmentally Preferable:
Specifications
About this item
Indicator-free version of DNA-ExitusPlus™. Decontamination solution for the removal of DNA and RNA contaminations from surfaces.
Delivery: Supplied in spray bottles.