N-L-alpha-Aspartyl-L-phenylalanine methyl ester
Catalog # APOSOR350295-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:100 g
- Protein synonyms:Aspartame
- Protein/peptide name:N-L-alpha-Aspartyl-L-phenylalanine Methyl Ester
- Purity:0.98