Human Recombinant B2 Microglobin (from E. coli)
Supplier: Apollo Scientific
Certificates
About this item
Recombinant HuB2M produced in E. coli, sterile clear liquid, 1mg/mg, >95% (HPLC, SDS-PAGE)
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein/peptide name:B2 Microglobin
Specifications
Frequently Bought Together