0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Proteins and Peptides
Recombinant HTLV-1 p24 Core
Recombinant HTLV-1 p24 Core
APOSBITP1909-1MG
:
Apollo Scientific
:
Recombinant HTLV-1 p24 Core
APOSBITP1909-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HTLV-1 p24c
Protein/peptide name:
HTLV-1 p24 Core