Order Entry
Belgium
ContactUsLinkComponent
Recombinant HIV-2 Envelope
Recombinant HIV-2 Envelope
Catalog # APOSBITP1906-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant HIV-2 Envelope
Catalog # APOSBITP1906-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
  • Species:
  • Environmentally Preferable:
  • Protein/peptide name:
    HIV-2 Envelope

Specifications

About this item