Recombinant Treponema pallidum p17 (partial)
Catalog # APOSBITP1856-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:
- Species:
- Environmentally Preferable:
- Protein synonyms:Treponema p17
- Protein/peptide name:Treponema pallidum p17 (partial)