Order Entry
Belgium
ContactUsLinkComponent
Recombinant Hepatitis C Virus NS5, HRP (Horseradish Peroxidase)
Recombinant Hepatitis C Virus NS5, HRP (Horseradish Peroxidase)
Catalog # APOSBITP1810-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis C Virus NS5, HRP (Horseradish Peroxidase)
Catalog # APOSBITP1810-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HCVNS5
  • Protein/peptide name:
    Hepatitis C Virus NS5

Specifications

About this item