Recombinant Hepatitis C Virus NS5 Genotype-5
Catalog # APOSBITP1805-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Protein synonyms:HCV NS5 Genotype-5
- Protein/peptide name:Hepatitis C Virus NS5 Genotype-5