Recombinant Hepatitis C Virus NS4, HRP (Horseradish Peroxidase)
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Conjugation:HRP (Horseradish Peroxidase)
- Protein/peptide type:Recombinant
- Protein synonyms:HCV NS4
- Protein/peptide name:Hepatitis C Virus NS4
Specifications
Frequently Bought Together