Order Entry
Belgium
ContactUsLinkComponent
Recombinant Chlamydia Trachomatis HSP70 (549-660) (from CHO cells)
Recombinant Chlamydia Trachomatis HSP70 (549-660) (from CHO cells)
Catalog # APOSBITP1744-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Chlamydia Trachomatis HSP70 (549-660) (from CHO cells)
Catalog # APOSBITP1744-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Protein synonyms:
    Chlamydia HSP70
  • Protein/peptide name:
    Chlamydia Trachomatis HSP70 (549-660)
  • Sequence:
    549-660

Specifications

About this item