Order Entry
Belgium
ContactUsLinkComponent
Recombinant Phospholipase A2 P00630 Bee Venom Protein
Recombinant Phospholipase A2 P00630 Bee Venom Protein
Catalog # APOSBITP1721-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Phospholipase A2 P00630 Bee Venom Protein
Catalog # APOSBITP1721-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Source:
  • Species:
  • Environmentally Preferable:
  • Protein synonyms:
    PA2-BVP
  • Protein/peptide name:
    Phospholipase A2 P00630 Bee Venom Protein

Specifications

About this item