Human recombinant zinc-alpha 2 glycoprotein
Catalog # APOSBITP1695-10UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:ZA2G
- Protein/peptide name:Zinc-Alpha 2 Glycoprotein