Order Entry
Belgium
ContactUsLinkComponent
Human recombinant ubiquitin Lys 48 to Arg 48 mutation
Human recombinant ubiquitin Lys 48 to Arg 48 mutation
Catalog # APOSBITP1683-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant ubiquitin Lys 48 to Arg 48 mutation
Catalog # APOSBITP1683-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Ubiquitin K48R
  • Protein/peptide name:
    Ubiquitin Lys 48 to Arg 48 mutation

Specifications

About this item