Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant S100A4
Human recombinant S100A4
Human recombinant S100A4
APOSBITP1643-5UG
:
Apollo Scientific
:
Human recombinant S100A4
APOSBITP1643-5UG
:
Apollo Scientific
:
Pk:
5 µG
Protein/peptide type:
Recombinant
Species:
Human
Protein/peptide name:
S100A4