Order Entry
Belgium
ContactUsLinkComponent
Human recombinant endostatin
Human recombinant endostatin
Catalog # APOSBITP1539-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant endostatin
Catalog # APOSBITP1539-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    Endostatin

Specifications

About this item