Order Entry
Belgium
ContactUsLinkComponent
Human recombinant CD14 (from CHO cells)
Human recombinant CD14 (from CHO cells)
Catalog # APOSBITP1492-10UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant CD14 (from CHO cells)
Catalog # APOSBITP1492-10UG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Protein/peptide name:
    CD14

Specifications

About this item