Human Recombinant Angiostatin Kringles (1-3)
Catalog # APOSBITP1451-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Angiostatin K1-3
- Protein/peptide name:Angiostatin Kringles (1-3)
- Sequence:1-3