Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant Tissue Plasminogen Activator
Human Recombinant Tissue Plasminogen Activator
Catalog # APOSBITP1356-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human Recombinant Tissue Plasminogen Activator
Catalog # APOSBITP1356-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    23
  • Protein synonyms:
    tPA
  • Protein/peptide name:
    Tissue Plasminogen Activator

Specifications

About this item