0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Proteins and Peptides
Denis recombinant GH
Denis recombinant GH
APOSBITP1078-1MG
:
Apollo Scientific
:
Denis recombinant GH
APOSBITP1078-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Denis
Protein synonyms:
Growth Hormone
Protein/peptide name:
GH