Order Entry
Belgium
ContactUsLinkComponent
Human recombinant GH Pit-20K
Human recombinant GH Pit-20K
Catalog # APOSBITP1068-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant GH Pit-20K
Catalog # APOSBITP1068-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Pituitary Growth Hormone-20K
  • Protein/peptide name:
    GH Pit-20K

Specifications

About this item