Human recombinant G-CSF (from CHO cells)
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Type de protéine/peptide:Recombinant
- Source:CHO cells
- Species:Human
- Synonyme de protéine:Granulocyte-Colony Stimulating Factor, derived
- Nom de la protéine/du peptide:G-CSF
Specifications
Frequently Bought Together