Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Mouse recombinant Acrp30
Mouse recombinant Acrp30
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Source:
Species:
Mouse
Environmentally Preferable:
Protein synonyms:
Adiponectin
Protein/peptide name:
Acrp30
Specifications
About this item
Product Details
Product Details