0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Proteins and Peptides
Recombinant exendin-4
Recombinant exendin-4
APOSBITH3013-1MG
:
Apollo Scientific
:
Recombinant exendin-4
APOSBITH3013-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Source:
Species:
Environmentally Preferable:
Protein/peptide name:
Exendin-4