Mouse recombinant MCP-3
Catalog # APOSBITC2029-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:
- Species:Mouse
- Environmentally Preferable:
- Protein synonyms:Monocyte Chemotactic Protein-3, CCL7
- Protein/peptide name:MCP-3