Order Entry
Belgium
ContactUsLinkComponent
Human recombinant MCP-3
Human recombinant MCP-3
Catalog # APOSBITC2028-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant MCP-3
Catalog # APOSBITC2028-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Monocyte Chemotactic Protein-3, CCL7
  • Protein/peptide name:
    MCP-3

Specifications

About this item