Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
2,3,3-Trimethyl-5-methoxy-3H-indole
2,3,3-Trimethyl-5-methoxy-3H-indole
2,3,3-Trimethyl-5-methoxy-3H-indole
Catalog # APOSBIB6278-1G
Supplier:
Apollo Scientific
CAS Number:
Expand image
2,3,3-Trimethyl-5-methoxy-3H-indole
Catalog # APOSBIB6278-1G
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 g
Seq:
70
Protein/peptide name:
2,3,3-Trimethyl-5-methoxy-3H-indole
Specifications
About this item
Product Details
Product Details