Orders LinkContactUsLinkComponent
Autoclavable muff covers, with Tyvek® drawstring, BHD
Autoclavable muff covers, with Tyvek® drawstring, BHD
Autoclavable muff covers, with Tyvek® drawstring, BHD
Autoclavable muff covers, with Tyvek® drawstring, BHD
Save time and standardise the activities in equipment preparation for sterilisation.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • Tear- and puncture-resistant
  • Significant process improvement compared to wrapping with sheets and tape
  • Tyvek® drawstring closure
BHD Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilisation to reduce time consuming wrapping of open containers. Each closure incorporates a Tyvek® drawstring closure to create the tightest possible seal around all openings.
 : Certificates of conformance sent with each shipment.
 : Conveniently packaged - 100 per sealed poly bag.
Product Family Options
  • Description
    Colour
  • BHD muff cover with Tyvek® drawstring, 5" (127 mm), fits 1,75 - 2,5" (44,5 - 63,5 mm) Ø
    Blue
    1 * 1.000 ST
    1 * 1.000 ST - 4 800,00 €
    4 800,00 €
    1 * 1.000 ST
       4 800,00 €
  • BHD muff cover with Tyvek® drawstring, 7" (178 mm), fits 2,5 - 4" (63 - 102 mm) Ø
    Blue
    1 * 1.000 ST
    1 * 1.000 ST - 5 270,00 €
    5 270,00 €
    1 * 1.000 ST
       5 270,00 €
  • BHD muff cover with Tyvek® drawstring, 9" (229 mm), fits 4 - 5,5" (102 - 140 mm) Ø
    Blue
    1 * 1.000 ST
    1 * 1.000 ST - 6 640,00 €
    6 640,00 €
    1 * 1.000 ST
       6 640,00 €
  • BHD muff cover with Tyvek® drawstring, 12" (305 mm), fits 5,5 - 8" (140 - 203 mm) Ø
    Blue
    1 * 1.000 ST
    1 * 1.000 ST - 7 530,00 €
    7 530,00 €
    1 * 1.000 ST
       7 530,00 €
  • BHD muff cover with Tyvek® drawstring, 16" (406 mm), fits 8 - 12" (203 - 305 mm) Ø
    Blue
    1 * 500 ST
    1 * 500 ST - 4 690,00 €
    4 690,00 €
    1 * 500 ST
       4 690,00 €