BHD Autoclavable Muff Covers, with Elastic
Supplier: KEYSTONE CLEANROOM PRODUCTS
Certificates
About this item
Save time and standardise activities in equipment preparation for sterilisation.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Simplifies conformance when wrapping to SOP's
- Steam sterilisation indicator - autoclavable up to 135 °C
- Tear and puncture resistant
- Significant process improvement compared to wrapping with sheets and tape
BHD muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilisation to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings.
Some items feature a 2" (51 mm) extension skirt, to allow removal of the muff without inadvertently touching the critical surfaces.
Delivery information: Certificates of Conformance sent with each shipment.
Packaging: Conveniently packed in 100 per sealed poly bag.
Frequently Bought Together