Order Entry
Belgium
ContactUsLinkComponent
 
NANOCOLOR® Photometric Tube Tests, Thiocyanate
Catalog # MANA985091
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Thiocyanate
Catalog # MANA985091
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Iron(III) -thiocyanate
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Thiocyanate 50, test no. 0-91
  • Dimensions:
    160×110×119 mm (Including packaging)
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    24
  • Weight:
    363 g (Including packaging)
  • For unit:
    UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
  • Tests per kit:
    20
  • Range:
    0,5 - 50,0 mg/L (SCN⁻)

Specifications

About this item

Photometric test kit for the determination of thiocyanate in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (AD) round cells. It is pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: Yes

Remark: Measuring range on NANOCOLOR VIS II

Hazardous material: Yes

Sludge reagent set: No

Digestion set: No

NanOx Metal: No

NanOx N: No

Delivery information:

Rugged box with 20 test tubes. Sufficient for 20 tests.