Silicate CRM, Standard Solutions, Supelco®
Supplier: Merck
Certificates
About this item
SiO₂ in H₂O
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST and USP
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Certifications: Traceable to SRM from NIST and USP.
Specifications
- M-Clarity quality level:MQ200