Order Entry
Belgium
ContactUsLinkComponent
Aluminium CRM, Standard Solutions, Supelco®
Aluminium CRM, Standard Solutions, Supelco®
Catalog # 1.32225.0100
Supplier:  Merck
CAS Number:  
Aluminium CRM, Standard Solutions, Supelco®
Catalog # 1.32225.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0,200 mg/L Al
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2
  • M-Clarity quality level:
    MQ200
  • Accuracy:
    ± 0,006 mg/L Al
  • pH range:
    3 - 5
  • For test kit(s):
    1.14825; 1.00594
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item

Al(NO₃)₃ in H₂O

  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors

Method used for Uncertainty Measurement (Accuracy in the table below)= ICP-OES

Certifications: Traceable to SRM from NIST and USP.