Order Entry
Belgium
ContactUsLinkComponent
2’-Deoxy-2’-fluoroadenosine-3’-CE-phosphoramidite ≥95%
2’-Deoxy-2’-fluoroadenosine-3’-CE-phosphoramidite ≥95%
Catalog # APOSBIMN2018-1G
Supplier:  Apollo Scientific
CAS Number:  
2’-Deoxy-2’-fluoroadenosine-3’-CE-phosphoramidite ≥95%
Catalog # APOSBIMN2018-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient