2’-Deoxy-2’-fluoroguanosine-3’-CE-phosphoramidite ≥95%
Catalog # APOSBIMN2017-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
Storage Temperature:Ambient