Capillary connection, b.safe
3,3 mm, 35 mm, 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm
Catalog # 590-0196
Supplier: Bohlender
Jump to:
Product Details & Documents
Straight fitting made of PTFE, capillary connection on both sides (UNF ¹/₄" thread), with 2 PFA fittings each for tubing Ext.Ø 1,6/2,2/3,2 mm.
- Excellent chemical resistance
- Available for a wide range of hose diameters
- The conical shape gets the connection tight and leak-free
Adaptor for connecting capillaries of the same size.
Specifications
- Hole Ø:3,3 mm
- With:2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm
- Length:35 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



