ContactUsLinkComponent
b.safe angled adaptor for canisters
b.safe angled adaptor for canisters
Catalog # 590-0333
Supplier:  Bohlender
undefined
b.safe angled adaptor for canisters
Catalog # 590-0333
Supplier:  Bohlender
Specifications
  • Description:
    Static dissipative angled adaptor for canisters, for vertical adjustment of b.safe Funnels 130 with thread S60 on b.safe canisters S60 with inclined neck
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
For vertical installation of b.safe funnels 130/180 on canister with inclined connectors and S60 thread. The insert made of static dissipative PE-EX is freely turnable inside the S60 screw cap (PPS) and thus allows vertical alignment of the funnel on the canister.
  • Stay flexible and independent
  • With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.