ContactUsLinkComponent
Mounting key for fittings, b.safe
  590-0215
 :  Bohlender
undefined
Mounting key for fittings, b.safe
  590-0215
 :  Bohlender
  • Description:
    Mounting key for b.safe fittings Ext.Ø. 1,6, 2,2 and 3,2 mm
  • Environmentally Preferable:
  • Pk:
    1
Inner shape fits the outer shape of b.safe fittings for capillary tubing.
  • Lateral slot to lead through tubing
  • With gripping surface
  • For tighten or loosen b.safe Fittings on b.safe caps and waste caps
Tightening: Put the fitting onto the tubing and position it in the requested connector. Lead in the tubing on the requested height. Put the mounting key and tighten the fitting. For disassembly, just put the Mounting Key onto the tubing and the fitting that is to be loosened, turn and you are ready.