Quick couplings, b.safe
About this item
Two-part tubing coupling made of PP with PFA Fitting to connect capillaries with Ext.Ø. 3,2 mm. The passage with the integrated quick lock is tightly closed through loosening the coupling. Tubing connections are easily interrupted without dripping from the tube, tubes and removal flasks are tightly closed during exchange. Version and scope of delivery as per below chart.
- Tightly closed
- The valves automatically open and a leakage free connection is now achieved
- If the parts are separated, the valves are automatically closed in order to avoid solvent escape
The capillary coupling is stacked together simply by hand. The coupling is successfully snapped if a clicking noise is audible. Depending on the combination of the coupling parts, the coupling can also be screwed as a capillary connection onto b.safe caps or just be used as quick connector of individual tubes.
5 Options Available Below
Product Details & Documents
Two-part tubing coupling made of PP with PFA Fitting to connect capillaries with Ext.Ø. 3,2 mm. The passage with the integrated quick lock is tightly closed through loosening the coupling. Tubing connections are easily interrupted without dripping from the tube, tubes and removal flasks are tightly closed during exchange. Version and scope of delivery as per below chart.
- Tightly closed
- The valves automatically open and a leakage free connection is now achieved
- If the parts are separated, the valves are automatically closed in order to avoid solvent escape
The capillary coupling is stacked together simply by hand. The coupling is successfully snapped if a clicking noise is audible. Depending on the combination of the coupling parts, the coupling can also be screwed as a capillary connection onto b.safe caps or just be used as quick connector of individual tubes.
Product Family Options
Quick coupling
Product Information
- ForAvailabilityPrice
- 590-0213Quick coupling, 2× UNF ¹/₄" (female), 2× fittingQuick coupling, 2× UNF ¹/₄" (female), 2× fittingTubing Ext.Ø: 3,2 mmLog in for availability1 * 1 ST119,00 €
- 590-0214Quick coupling, 1× UNF ¹/₄" (female), 1× UNF ¹/₄" (male), 1× fittingQuick coupling, 1× UNF ¹/₄" (female), 1× UNF ¹/₄" (male), 1× fittingTubing Ext.Ø: 3,2 mmLog in for availability1 * 1 ST120,00 €
Coupling pieces
Product Information
- ForAvailabilityPrice
- 590-0210Coupling piece, 1× UNF ¹/₄" (female) with coupling connection female, 1× fittingCoupling piece, 1× UNF ¹/₄" (female) with coupling connection female, 1× fittingTubing Ext.Ø: 3,2 mmLog in for availability1 * 1 ST72,50 €
- 590-0211Coupling piece, 1× UNF ¹/₄" (female) with coupling connection male, 1× fittingCoupling piece, 1× UNF ¹/₄" (female) with coupling connection male, 1× fittingTubing Ext.Ø: 3,2 mmLog in for availability1 * 1 ST50,80 €
- 590-0212Coupling piece, 1× UNF ¹/₄" (male) with coupling connection maleCoupling piece, 1× UNF ¹/₄" (male) with coupling connection maleTubing Ext.Ø: 3,2 mmLog in for availability1 * 1 ST55,60 €