Order Entry
Belgium
ContactUsLinkComponent
VWR®, GC Rich TEMPase Hot start DNA polymerase and MasterMix
VWR®, GC Rich TEMPase Hot start DNA polymerase and MasterMix
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, GC Rich TEMPase Hot start DNA polymerase and MasterMix
Supplier:  VWR Chemicals

Specifications

  • Application:
    Multiplex
  • Format:
    Enzyme with/without buffer
  • High fidelity:
    No
  • Hot start:
    Chemically modified
  • Storage conditions:
    – 20 °C (long term), + 4 °C (6 months)
  • Amplicon size:
    ≤ 5kb
  • Processivity:
    Standard
  • Exonuclease activity:
    5’→3’
  • Product overhang:
    3'A
  • Buffer composition:
    4x GC Buffer I, 4 x GC Buffer II and 25 mM MgCl2
  • Concentration:
    5 U/µl
  • Description:
    GC-rich DNA Target Kit
    TEMPase HotStart DNA Polymerase 5U/µl 500 units
  • Size:
    1 x 0.1 ml + 3 x 1.5 ml
  • Units:
    500
  • Features:
    GC-rich

Specifications

Frequently Bought Together