4-(Trifluoromethyl)phenyl isothiocyanate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1645-65-4
:
Formula:C₈H₄F₃NS
MDL Number:MFCD00039645
:203.19 g/mol
Boiling Pt:207 °C
Melting Pt:39…43 °C
Flash Pt:>110 °C
UN:3261
ADR:8,II
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together