
3-(Trifluoromethyl)-DL-phenylalanine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:63701-37-1
:
Formula:C₁₀H₁₀F₃NO₂
MDL Number:MFCD00069074
:233.19 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,II
:GHS07
Frequently Bought Together